3-Fluoro-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
F0670-25G F0670-5G
TCIAF0670-25GEA 3100 NOK
TCIAF0670-25G TCIAF0670-5G
3-Fluoro-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Fluoro-4-nitrobenzoic acid
Formel: C₇H₄FNO₄
MW: 185,11 g/mol
MDL nummer: MFCD01862092
CAS-nummer: 403-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a4dccdb1c1797188633a2bcdda90ebc9cf823c&vid=63a4dccdb1c1797188633a2bcdda90ebc9cf823c&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C