2,4-Difluorothiophenol ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,3-Difluoro-4-mercaptobenzene, 2,4-Difluorobenzenethiol, 2,4-Difluorobenzene-1-thiol

D3342-25G D3342-5G
TCIAD3342-25GEA 9400 NOK
TCIAD3342-25G TCIAD3342-5G
2,4-Difluorothiophenol ≥97.0% (by GC)
2,4-Difluorothiophenol
Formel: C₆H₄F₂S
MW: 146,16 g/mol
Kokepunkt: 59°C
Tetthet: 1,29 g/cm³ (20 °C)
Flash punkt: 49°C
MDL nummer: MFCD00203477
CAS-nummer: 1996-44-7
FN: 3336
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bc8340725a96a8c95a074b65bf2bdbeb553e554&vid=5bc8340725a96a8c95a074b65bf2bdbeb553e554&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR