1,12-Diaminododecane ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: N12N, Dodecamethylendiamine, 1,12-Dodecanediamine

D0091-25G D0091-500G D0091-100G
TCIAD0091-25GEA 730 NOK
TCIAD0091-25G TCIAD0091-500G TCIAD0091-100G
1,12-Diaminododecane ≥98.0% (by GC, titration analysis)
1,12-Diaminododecane
Formel: C₁₂H₂₈N₂
MW: 200,37 g/mol
Kokepunkt: 145°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008155
CAS-nummer: 2783-17-7
EINECS: 220-489-6
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d62b49709ff6862a275b92f1d650ed376d90029&vid=3d62b49709ff6862a275b92f1d650ed376d90029&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR