Biphenyl-3-ol ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 3-Phenylphenol, [1,1'-Biphenyl]-3-ol, 3-Hydroxybiphenyl, 3-Hydroxydiphenyl

P0768-25G P0768-5G
TCIAP0768-25GEA 3825 NOK
TCIAP0768-25G TCIAP0768-5G
Biphenyl-3-ol ≥97.0% (by GC)
Biphenyl-3-ol
Formel: C₁₂H₁₀O
MW: 170,21 g/mol
Kokepunkt: 334 °C (1013 hPa)
Smeltepunkt: 78 °C
MDL nummer: MFCD00002294
CAS-nummer: 580-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd1bac67896aa780be5e64f40d36de1da3501f19&vid=fd1bac67896aa780be5e64f40d36de1da3501f19&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 75.0 to 81.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR