(2-Fluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid

F0205-25G F0205-5G
TCIAF0205-25GEA 865 NOK
TCIAF0205-25G TCIAF0205-5G
(2-Fluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Fluorophenyl)acetic acid
Formel: (C₆H₄F)CH₂COOH
MW: 154,14 g/mol
Kokepunkt: 263 °C (1013 hPa)
Smeltepunkt: 64 °C
MDL nummer: MFCD00004315
CAS-nummer: 451-82-1
EINECS: 207-196-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e2f0d4f744b0fb7486d65fdcfbc5d141d0a72fc&vid=9e2f0d4f744b0fb7486d65fdcfbc5d141d0a72fc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 63.0 to 66.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR