1-Phenyl-2-propanol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
(2-Hydroxypropyl)benzol, (±)-1-Phenyl-2-propanol, (±)-α-Methylphenethyl alcohol, Benzylmethylcarbinol
P0215-25G
P0215-5G
TCIAP0215-25GEA
1550
NOK
TCIAP0215-25G
TCIAP0215-5G
1-Phenyl-2-propanol ≥98.0% (by GC)
1-Phenyl-2-propanol
Formel:
C₆H₅CH₂CH(OH)CH₃ MW: 136,19 g/mol Kokepunkt: 213°C Smeltepunkt: 65…67 °C Tetthet: 0,997 g/cm³ (20 °C) Flash punkt: 85°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004540 CAS-nummer: 698-87-3 EINECS: 211-821-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...