(3,4,5-Trimethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 3,4,5-Trimethoxyphenylacetic acid
T1105-25G T1105-5G
TCIAT1105-25GEA 2000 NOK
TCIAT1105-25G TCIAT1105-5G
(3,4,5-Trimethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3,4,5-Trimethoxyphenyl)acetic acid
Formel: (C₆H₂(OCH₃)₃)CH₂COOH
MW: 226,23 g/mol
Kokepunkt: 352 °C (1013 hPa)
Smeltepunkt: 117…120 °C
MDL nummer: MFCD00004336
CAS-nummer: 951-82-6
EINECS: 213-456-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71ecea83a5135c8bca4aa76d8aa1f24f455ee0de&vid=71ecea83a5135c8bca4aa76d8aa1f24f455ee0de&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C
Solubility in Methanol almost transparency