(3,5-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3,5-Difluorophenylacetic acid

D3462-1G D3462-5G
TCIAD3462-1GEA 285 NOK
TCIAD3462-1G TCIAD3462-5G
(3,5-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3,5-Difluorophenyl)acetic acid
Formel: (C₆H₃F₂)CH₂COOH
MW: 172,13 g/mol
MDL nummer: MFCD00010316
CAS-nummer: 105184-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db5547c2a354dd50bd6b5ccf823d73d23277adb8&vid=db5547c2a354dd50bd6b5ccf823d73d23277adb8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 67.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR