(α,α,α-Trifluoro-3-tolyl)acetic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: (3-(Trifluoromethyl)phenyl)acetic acid, 3-(Trifluoromethyl)phenylacetic acid

T1662-25G T1662-5G
TCIAT1662-25GEA 1650 NOK
TCIAT1662-25G TCIAT1662-5G
(α,α,α-Trifluoro-3-tolyl)acetic acid ≥98.0% (by titrimetric analysis)
(α,α,α-Trifluoro-3-tolyl)acetic acid
Formel: (C₆H₄(CF₃))CH₂COOH
MW: 204,15 g/mol
MDL nummer: MFCD00004339
CAS-nummer: 351-35-9
EINECS: 206-511-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f99eab6b9545e3842b4fdd54d609a3fd85267c7&vid=4f99eab6b9545e3842b4fdd54d609a3fd85267c7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 76.0 to 79.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR