2',4'-Dihydroxyacetophenone ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
1-(2,4-Dihydroxyphenyl)ethan-1-one, 2,4-Dihydroxyacetophenone
D0561-25G
D0561-500G
D0561-100G
TCIAD0561-25GEA
365
NOK
TCIAD0561-25G
TCIAD0561-500G
TCIAD0561-100G
2',4'-Dihydroxyacetophenone ≥98.0% (by GC, titration analysis)
2',4'-Dihydroxyacetophenone
Formel:
C₈H₈O₃ MW: 152,15 g/mol Kokepunkt: 336 °C (1013 hPa) Smeltepunkt: 142…143 °C Tetthet: 1,18 g/cm³ (141 °C) |
MDL nummer:
MFCD00002279 CAS-nummer: 89-84-9 EINECS: 201-945-3 Merck Indeks: 14,08140 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 144.0 to 147.0 deg-C |
Solubility in Methanol | faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...