3',5'-Dihydroxyacetophenone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone
D1981-10G
D1981-25G
TCIAD1981-10GEA
745
NOK
TCIAD1981-10G
TCIAD1981-25G
3',5'-Dihydroxyacetophenone ≥98.0% (by GC)
3',5'-Dihydroxyacetophenone
Formel:
C₈H₈O₃ MW: 152,15 g/mol Kokepunkt: 296 °C (1013 hPa) Smeltepunkt: 148 °C |
MDL nummer:
MFCD00002290 CAS-nummer: 51863-60-6 EINECS: 257-480-1 |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...