Dietylmalonat ≥99.0% (by GC)

Leverandør: TCI

Synonymer: 1,3-Propanedioic acid diethyl ester, Malonic acid diethyl ester

M0029-25ML M0029-500ML
TCIAM0029-25MLEA 220 NOK
TCIAM0029-25ML TCIAM0029-500ML
Dietylmalonat ≥99.0% (by GC)
Dietylmalonat
Formel: C₇H₁₂O₄
MW: 160,17 g/mol
Kokepunkt: 199°C
Smeltepunkt: -50°C
Tetthet: 1,055 g/cm³ (20 °C)
Flash punkt: 93°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009195
CAS-nummer: 105-53-3
EINECS: 203-305-9
Merck Indeks: 14,03823
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49dcbb51cbfda51dd1ef18b027d9c9288b9b2144&vid=49dcbb51cbfda51dd1ef18b027d9c9288b9b2144&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR