2',5'-Dimethoxyacetophenone ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1-(2,5-Dimethoxyphenyl)ethan-1-one, 1-(2,5-Dimethoxyphenyl)ethanone

D1444-25G D1444-500G
TCIAD1444-25GEA 570 NOK
TCIAD1444-25G TCIAD1444-500G
2',5'-Dimethoxyacetophenone ≥98.0% (by GC)
2',5'-Dimethoxyacetophenone
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Kokepunkt: 158°C
Smeltepunkt: 18…20 °C
Tetthet: 1,139 g/cm³ (20 °C)
MDL nummer: MFCD00008728
CAS-nummer: 1201-38-3
EINECS: 214-858-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a258c277083fdecf9c18a95ebbd41056798fc00&vid=1a258c277083fdecf9c18a95ebbd41056798fc00&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR