1,6-Heptadiyne ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
H0483-5ML
H0483-1ML
TCIAH0483-5MLEA
1025
NOK
TCIAH0483-5ML
TCIAH0483-1ML
1,6-Heptadiyne ≥98.0% (by GC)
1,6-Heptadiyne
Formel:
C₇H₈ MW: 92,14 g/mol Kokepunkt: 112°C Smeltepunkt: –85 °C Tetthet: 0,8164 g/cm³ (17 °C) Flash punkt: 9°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00014925 CAS-nummer: 2396-63-6 EINECS: 219-253-5 FN: 3295 ADR: 3,II |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...