Dimethyl suberate ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Dimethyl octanedioate, 1,8-Octanedioic acid dimethyl ester, Suberic acid dimethyl ester
O0183-25ML
O0183-500ML
TCIAO0183-25MLEA
1750
NOK
TCIAO0183-25ML
TCIAO0183-500ML
Dimethyl suberate ≥98.0% (by GC)
Dimethyl suberate
Formel:
C₁₀H₁₈O₄ MW: 202,25 g/mol Kokepunkt: 270°C Smeltepunkt: –3,1 °C Tetthet: 1,014 g/cm³ (18 °C) |
MDL nummer:
MFCD00008471 CAS-nummer: 1732-09-8 EINECS: 217-059-5 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...