N,N-Dimethyldodecylamine ≥96.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: N,n-dimethyldodecylamine, Dodecyldimethylamine

D0002-25ML D0002-500ML
TCIAD0002-25MLEA 230 NOK
TCIAD0002-25ML TCIAD0002-500ML
N,N-Dimethyldodecylamine ≥96.0% (by GC, titration analysis)
N,N-Dimethyldodecylamine
Formel: C₁₄H₃₁N
MW: 213,41 g/mol
Kokepunkt: 149°C
Smeltepunkt: –20 °C
Tetthet: 0,787 g/cm³ (20 °C)
Flash punkt: 130°C
MDL nummer: MFCD00008970
CAS-nummer: 112-18-5
EINECS: 203-943-8
FN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=242e701f81c09aaf6f648b70319617e6a88087f9&vid=242e701f81c09aaf6f648b70319617e6a88087f9&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR