2-Fluoro-p-toluidine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Danger

F0529-25G F0529-5G
TCIAF0529-25GEA 2075 NOK
TCIAF0529-25G TCIAF0529-5G
2-Fluoro-p-toluidine ≥97.0% (by GC, titration analysis)
2-Fluoro-p-toluidine
Formel: C₇H₈FN
Kokepunkt: 71°C
Smeltepunkt: 3°C
Tetthet: 1,11
Flash punkt: 79°C
MDL nummer: MFCD00040975
CAS-nummer: 452-80-2
FN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57f3fa10dc3bbf730a4237a76e169d4cd0d875d0&vid=57f3fa10dc3bbf730a4237a76e169d4cd0d875d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR