4-Nitro-m-toluidine ≥97.0% (by GC)

Leverandør: TCI
Danger

M1677-25G M1677-5G
TCIAM1677-25GEA 2125 NOK
TCIAM1677-25G TCIAM1677-5G
4-Nitro-m-toluidine ≥97.0% (by GC)
4-Nitro-m-toluidine
Formel: C₇H₈N₂O₂
Smeltepunkt: 138 °C
MDL nummer: MFCD00091833
CAS-nummer: 611-05-2
EINECS: 210-247-8
FN: 2660
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a03240f23e20163890b7c6e5d84d191a1f3049&vid=62a03240f23e20163890b7c6e5d84d191a1f3049&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 131.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR