4-Fluoro-3-(trifluoromethyl)aniline ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Amino-2-fluorobenzotrifluoride, 5-Amino-α,α,α,2-tetrafluorotoluene, α,α,α,4-Tetrafluoro-m-toluidine, 5-Amino-alpha,alpha,alpha,2-tetrafluorotoluene

A1164-25G A1164-5G
TCIAA1164-25GEA 1450 NOK
TCIAA1164-25G TCIAA1164-5G
4-Fluoro-3-(trifluoromethyl)aniline ≥98.0% (by GC)
4-Fluoro-3-(trifluoromethyl)aniline
Formel: C₇H₅F₄N
MW: 179,12 g/mol
Kokepunkt: 208°C
Tetthet: 1,39 g/cm³ (20 °C)
Flash punkt: 92°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007834
CAS-nummer: 2357-47-3
EINECS: 219-095-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5905932401ceacbef81fc500a3306cb252f0f0f5&vid=5905932401ceacbef81fc500a3306cb252f0f0f5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR