1-Bromo-2-chloro-5-fluorobenzene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Bromo-1-chloro-4-fluorobenzene
B3048-25G
B3048-5G
TCIAB3048-25GEA
2050
NOK
TCIAB3048-25G
TCIAB3048-5G
1-Bromo-2-chloro-5-fluorobenzene ≥98.0% (by GC)
1-Bromo-2-chloro-5-fluorobenzene
Formel:
C₆H₃BrClF MW: 209,44 g/mol |
MDL nummer:
MFCD00061120 CAS-nummer: 201849-15-2 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...