3-Bromobenzyl alcohol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (3-Bromophenyl)methanol

B1287-25G B1287-5G
TCIAB1287-25GEA 1025 NOK
TCIAB1287-25G TCIAB1287-5G
3-Bromobenzyl alcohol ≥98.0% (by GC)
3-Bromobenzyl alcohol
Formel: C₇H₇BrO
MW: 187,04 g/mol
Kokepunkt: 165°C
Tetthet: 1,56 g/cm³ (20 °C)
MDL nummer: MFCD00004629
CAS-nummer: 15852-73-0
EINECS: 239-975-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR