1-Bromo-3-chloro-5-fluorobenzene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3-Chloro-5-fluorobromobenzene
B3045-25G
B3045-5G
TCIAB3045-25GEA
1050
NOK
TCIAB3045-25G
TCIAB3045-5G
1-Bromo-3-chloro-5-fluorobenzene ≥98.0% (by GC)
1-Bromo-3-chloro-5-fluorobenzene
Formel:
C₆H₃BrClF MW: 209,44 g/mol Kokepunkt: 72°C |
MDL nummer:
MFCD00070747 CAS-nummer: 33863-76-2 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...