2-Bromo-4-fluoroanisole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Bromo-4-fluoro-1-methoxybenzene

B1830-25G B1830-5G
TCIAB1830-25GEA 930 NOK
TCIAB1830-25G TCIAB1830-5G
2-Bromo-4-fluoroanisole ≥98.0% (by GC)
2-Bromo-4-fluoroanisole
Formel: C₇H₆BrFO
MW: 205,03 g/mol
Kokepunkt: 79°C
Smeltepunkt: –27 °C
Tetthet: 1,58 g/cm³ (20 °C)
Flash punkt: 98°C
MDL nummer: MFCD00012014
CAS-nummer: 452-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f0dfe5d474685e7bdf1646fa67ff5ed0e2f24e&vid=e9f0dfe5d474685e7bdf1646fa67ff5ed0e2f24e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR