4'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
|
Warning
|
H0193-25G
H0193-500G
H0193-100G
TCIAH0193-25GEA
220
NOK
TCIAH0193-25G
TCIAH0193-500G
TCIAH0193-100G
4'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
4'-Hydroxyacetophenone
Formel:
C₈H₈O₂ MW: 136,15 g/mol Kokepunkt: 148°C Smeltepunkt: 105…108 °C Tetthet: 1,139 g/cm³ (25 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00002359 CAS-nummer: 99-93-4 EINECS: 202-802-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 109.0 to 112.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...