4-Fluorobenzyl alcohol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (4-Fluorophenyl)methanol

F0251-25G F0251-5G
TCIAF0251-25GEA 835 NOK
TCIAF0251-25G TCIAF0251-5G
4-Fluorobenzyl alcohol ≥98.0% (by GC)
4-Fluorobenzyl alcohol
Formel: C₇H₇FO
MW: 126,13 g/mol
Kokepunkt: 99°C
Smeltepunkt: 22…23 °C
Tetthet: 1,15 g/cm³ (20 °C)
MDL nummer: MFCD00004651
CAS-nummer: 459-56-3
EINECS: 207-292-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35ac861f5562e6a5b88bf6bfb849b89e8c506e14&vid=35ac861f5562e6a5b88bf6bfb849b89e8c506e14&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR