2,4,6-Tribromoanisole ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
1,3,5-Tribromo-2-methoxybenzene
T2301-25G
T2301-5G
TCIAT2301-25GEA
1625
NOK
TCIAT2301-25G
TCIAT2301-5G
2,4,6-Tribromoanisole ≥98.0% (by GC)
2,4,6-Tribromoanisole
Formel:
C₇H₅Br₃O MW: 344,83 g/mol Kokepunkt: 299°C Smeltepunkt: 84…88 °C Tetthet: 2,13 g/cm³ (20 °C) |
MDL nummer:
MFCD00192510 CAS-nummer: 607-99-8 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...