2,4-Dichlorobenzoyl chloride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 2,4-Dichlorobenzene-1-carbonyl chloride

D0345-25G D0345-500G
TCIAD0345-25GEA 245 NOK
TCIAD0345-25G TCIAD0345-500G
2,4-Dichlorobenzoyl chloride ≥97.0% (by titrimetric analysis)
2,4-Dichlorobenzoyl chloride
Formel: C₇H₃Cl₃O
MW: 209,46 g/mol
Kokepunkt: 252 °C (1013 hPa)
Smeltepunkt: 18 °C
Tetthet: 1,49 g/cm³ (20 °C)
Flash punkt: 137°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000661
CAS-nummer: 89-75-8
EINECS: 201-936-4
FN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=142a3f926f885ac29b952c90ff27f50a70c03f02&vid=142a3f926f885ac29b952c90ff27f50a70c03f02&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (Argentmetric Titration) min. 97.0 %
Freezing point 19.0 to 22.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR