2-Chloro-5-fluoropyridine ≥98.0% (by GC)

Leverandør: TCI
Warning

C2270-1G C2270-5G
TCIAC2270-1GEA 245 NOK
TCIAC2270-1G TCIAC2270-5G
2-Chloro-5-fluoropyridine ≥98.0% (by GC)
2-Chloro-5-fluoropyridine
Formel: C₅H₃ClFN
MW: 131,54 g/mol
Kokepunkt: 74°C
Flash punkt: 42°C
MDL nummer: MFCD03095332
CAS-nummer: 31301-51-6
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc087f29f59e950760d0e6a9913e1896ad7f65ed&vid=dc087f29f59e950760d0e6a9913e1896ad7f65ed&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR