2-Ethyl-3-methylpyrazine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
E0361-25ML
E0361-5ML
TCIAE0361-25MLEA
660
NOK
TCIAE0361-25ML
TCIAE0361-5ML
2-Ethyl-3-methylpyrazine ≥98.0% (by GC, titration analysis)
2-Ethyl-3-methylpyrazine
Formel:
C₇H₁₀N₂ MW: 122,17 g/mol Kokepunkt: 70°C Tetthet: 0,99 g/cm³ (20 °C) Flash punkt: 57°C |
MDL nummer:
MFCD00006150 CAS-nummer: 15707-23-0 EINECS: 239-799-8 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...