Pyridine-3,4-dicarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3,4-Pyridinedicarboxylic acid, Cinchomeronic acid, Pyridine-3,4-dicarboxylicacid

P0682-25G P0682-5G
TCIAP0682-25GEA 1250 NOK
TCIAP0682-25G TCIAP0682-5G
Pyridine-3,4-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Pyridine-3,4-dicarboxylic acid
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Kokepunkt: 529 °C (1013 hPa)
Smeltepunkt: 260°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006392
CAS-nummer: 490-11-9
EINECS: 207-705-4
Merck Indeks: 14,02283
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67835f8ab42c67357a4d59e1ffb467eff4fb591d&vid=67835f8ab42c67357a4d59e1ffb467eff4fb591d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR