3,5-Dichlorobenzoyl chloride ≥95.0% (by GC)

Leverandør: TCI
Danger

D1404-25G D1404-500G
TCIAD1404-25GEA 340 NOK
TCIAD1404-25G TCIAD1404-500G
3,5-Dichlorobenzoyl chloride ≥95.0% (by GC)
3,5-Dichlorobenzoyl chloride
Formel: C₇H₃Cl₃O
MW: 209,46 g/mol
Kokepunkt: 249 °C (1013 hPa)
Smeltepunkt: 26…28 °C
Tetthet: 1,478 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009817
CAS-nummer: 2905-62-6
EINECS: 220-813-6
FN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd62dfdddb0c3f816f9073a00256168916ad46&vid=fabd62dfdddb0c3f816f9073a00256168916ad46&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 95.0 %
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR