3-Chloro-p-toluic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

C2462-25G C2462-5G
TCIAC2462-25GEA 1700 NOK
TCIAC2462-25G TCIAC2462-5G
3-Chloro-p-toluic acid ≥98.0% (by GC, titration analysis)
3-Chloro-p-toluic acid
Formel: C₈H₇ClO₂
MW: 170,6 g/mol
MDL nummer: MFCD00045841
CAS-nummer: 5162-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=892f32ff173709d84ffacb67627e80f57527c246&vid=892f32ff173709d84ffacb67627e80f57527c246&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 200.0 to 204.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR