4-Amino-3,5-dichloropyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3,5-Dichloro-4-pyridylamine, 3,5-Dichloropyridin-4-amine

A1545-25G A1545-5G
TCIAA1545-25GEA 890 NOK
TCIAA1545-25G TCIAA1545-5G
4-Amino-3,5-dichloropyridine ≥98.0% (by GC, titration analysis)
4-Amino-3,5-dichloropyridine
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
MDL nummer: MFCD00125023
CAS-nummer: 22889-78-7
EINECS: 245-304-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=757102df92c8f29e29243dad167de237ba06c792&vid=757102df92c8f29e29243dad167de237ba06c792&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 160.0 to 163.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR