4-Bromo-3-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
B3791-25G B3791-5G
TCIAB3791-25GEA 2325 NOK
TCIAB3791-25G TCIAB3791-5G
4-Bromo-3-chlorobenzoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-3-chlorobenzoic acid
Formel: C₇H₄BrClO₂
MW: 235,46 g/mol
MDL nummer: MFCD08276864
CAS-nummer: 25118-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86258ec4db1c99b9bf26f5be64611eb1ae42b133&vid=86258ec4db1c99b9bf26f5be64611eb1ae42b133&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 220.0 to 224.0 deg-C