Isonicotinonitrile ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Pyridinecarbonitrile, Pyridine-4-carbonitrile, 4-Cyanopyridine
C0457-25G
C0457-500G
C0457-100G
TCIAC0457-25GEA
220
NOK
TCIAC0457-25G
TCIAC0457-500G
TCIAC0457-100G
Isonicotinonitrile ≥98.0% (by GC, titration analysis)
Isonicotinonitrile
Formel:
C₆H₄N₂ MW: 104,11 g/mol Kokepunkt: 198°C Smeltepunkt: 78…80 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00006417 CAS-nummer: 100-48-1 EINECS: 202-856-2 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...