3-Methyl-2-buten-1-ol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Methyl-2-buten-4-ol

M0714-25ML M0714-500ML M0714-100ML
TCIAM0714-25MLEA 230 NOK
TCIAM0714-25ML TCIAM0714-500ML TCIAM0714-100ML
3-Methyl-2-buten-1-ol ≥98.0% (by GC)
3-Methyl-2-buten-1-ol
Formel: C₅H₁₀O
MW: 86,13 g/mol
Kokepunkt: 140°C
Smeltepunkt: –86 °C
Tetthet: 0,848 g/cm³ (20 °C)
Flash punkt: 53°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002916
CAS-nummer: 556-82-1
EINECS: 209-141-4
FN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f349b9e9b4ca9ed0b3ab1a8379163bd16de3ade2&vid=f349b9e9b4ca9ed0b3ab1a8379163bd16de3ade2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR