Indole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

I0722-1G I0722-5G
TCIAI0722-1GEA 1150 NOK
TCIAI0722-1G TCIAI0722-5G
Indole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Indole-4-carboxylic acid
Formel: C₉H₇NO₂
MW: 161.16 g/mol
MDL nummer: MFCD00009739
CAS-nummer: 2124-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f24d18788e0a7937c9d149bf5c86258792b74a1&vid=4f24d18788e0a7937c9d149bf5c86258792b74a1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 213.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR