4-Methylimidazole ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4-Methyl-1H-imidazole

M0636-25G M0636-500G M0636-100G
TCIAM0636-25GEA 260 NOK
TCIAM0636-25G TCIAM0636-500G TCIAM0636-100G
4-Methylimidazole ≥98.0% (by GC)
4-Methylimidazole
Formel: C₄H₆N₂
MW: 82,11 g/mol
Kokepunkt: 263°C
Tetthet: 1,0416 g/cm³ (14.3 °C)
MDL nummer: MFCD00005201
CAS-nummer: 822-36-6
EINECS: 212-497-3
FN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4329e38c72e5243831bf2d392af9be3f92504d07&vid=4329e38c72e5243831bf2d392af9be3f92504d07&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR