5-Bromovaleric acid ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 5-Bromopentanoic acid

B1169-10G B1169-25G
TCIAB1169-10GEA 245 NOK
TCIAB1169-10G TCIAB1169-25G
5-Bromovaleric acid ≥97.0% (by GC, titration analysis)
5-Bromovaleric acid
Formel: BrH₂C(CH₂)₃COOH
MW: 181,03 g/mol
Kokepunkt: 152°C
Smeltepunkt: 38…40 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004414
CAS-nummer: 2067-33-6
EINECS: 218-185-3
FN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a792ed3bc2f12df6a22f4debf778570ea194191f&vid=a792ed3bc2f12df6a22f4debf778570ea194191f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 39.0 to 43.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR