6-Methylindole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 6-Methyl-1H-indole

M1430-1G M1430-5G
TCIAM1430-1GEA 985 NOK
TCIAM1430-1G TCIAM1430-5G
6-Methylindole ≥98.0% (by GC)
6-Methylindole
Formel: C₉H₉N
MW: 131,18 g/mol
Kokepunkt: 260 °C (1013 hPa)
Smeltepunkt: 30…31 °C
Tetthet: 1,05 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005682
CAS-nummer: 3420-02-8
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=714f7a434d524f0e4886f35de7dfa70645049f61&vid=714f7a434d524f0e4886f35de7dfa70645049f61&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Freezing point gt. 25.0 to 29.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR