Dimethyl trisulphide ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Dimethyltrisulphide

D3418-25G D3418-5G
TCIAD3418-25GEA 1825 NOK
TCIAD3418-25G TCIAD3418-5G
Dimethyl trisulphide ≥98.0% (by GC)
Dimethyl trisulphide
Formel: C₂H₆S₃
MW: 126,27 g/mol
Kokepunkt: 170°C
Smeltepunkt: -68°C
Flash punkt: 56°C
MDL nummer: MFCD00039808
CAS-nummer: 3658-80-8
EINECS: 222-910-9
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7f54bf56b0d9049f17d986a80eb54fc75f53e81&vid=f7f54bf56b0d9049f17d986a80eb54fc75f53e81&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR