Dietyloksalat ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Oxalic acid diethyl ester

O0078-25G O0078-500G
TCIAO0078-25GEA 220 NOK
TCIAO0078-25G TCIAO0078-500G
Dietyloksalat ≥98.0% (by GC)
Dietyloksalat
Formel: C₆H₁₀O₄
MW: 146,14 g/mol
Kokepunkt: 185°C
Smeltepunkt: -41°C
Tetthet: 1,0785 g/cm³ (20 °C)
Flash punkt: 76°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009119
CAS-nummer: 95-92-1
EINECS: 202-464-1
FN: 2525
ADR: 6.1,III
Merck Indeks: 14,03125
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f71832de944518156dbf150c3e97dfea2b6376da&vid=f71832de944518156dbf150c3e97dfea2b6376da&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR