Dodecyl acetate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: Lauryl acetate, Acetic acid Lauryl ester, Acetic acid Dodecyl ester

A0902-25ML A0902-500ML
TCIAA0902-25MLEA 220 NOK
TCIAA0902-25ML TCIAA0902-500ML
Dodecyl acetate ≥95.0% (by GC)
Dodecyl acetate
Formel: C₁₄H₂₈O₂
MW: 228,38 g/mol
Kokepunkt: 180°C
Tetthet: 0,865 g/cm³ (25 °C)
Flash punkt: 140°C
MDL nummer: MFCD00008973
CAS-nummer: 112-66-3
EINECS: 203-995-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f06a926c20b99cf6ef1a3b9355428394b7f4abe2&vid=f06a926c20b99cf6ef1a3b9355428394b7f4abe2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 97.0 %
Acid Value max. 1.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR