N,N-Dibutylformamid ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Formdibutylamide, Formic acid dibutylamide

D2147-25ML D2147-500ML D2147-100ML
TCIAD2147-25MLEA 230 NOK
TCIAD2147-25ML TCIAD2147-500ML TCIAD2147-100ML
N,N-Dibutylformamid ≥98.0% (by GC)
N,N-Dibutylformamid
Formel: C₉H₁₉NO
MW: 157,26 g/mol
Kokepunkt: 243°C
Smeltepunkt: –30 °C
Tetthet: 0,8932 g/cm³ (20 °C)
Flash punkt: 100°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003289
CAS-nummer: 761-65-9
EINECS: 212-090-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94bcd753bad8a61fc4a307b09b54f6c86de4e36d&vid=94bcd753bad8a61fc4a307b09b54f6c86de4e36d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR