Stearoyl chloride ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
S0404-25G
S0404-250G
TCIAS0404-25GEA
970
NOK
TCIAS0404-25G
TCIAS0404-250G
Stearoyl chloride ≥97.0% (by GC, titration analysis)
Stearoyl chloride
Formel:
C₁₈H₃₅ClO MW: 302,93 g/mol Kokepunkt: 215°C Smeltepunkt: 164…167 °C Tetthet: 0,9007 g/cm³ (25 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000744 CAS-nummer: 112-76-5 EINECS: 204-004-5 FN: 3265 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...