Methyl propionate ≥99.0% (by GC)
Leverandør: TCI
Danger
Synonymer: Propanoic acid methyl ester
P0508-25ML P0508-500ML
TCIAP0508-25MLEA 220 NOK
TCIAP0508-25ML TCIAP0508-500ML
Methyl propionate ≥99.0% (by GC)
Methyl propionate
Formel: C₄H₈O₂
MW: 88,11 g/mol
Kokepunkt: 80°C
Smeltepunkt: -88°C
Tetthet: 0,91 g/cm³ (20 °C)
Flash punkt: -4°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009306
CAS-nummer: 554-12-1
EINECS: 209-060-4
FN: 1248
ADR: 3,II
Merck Indeks: 14,06112
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee64c0d09794e86969c8bf59d36644293bf59f88&vid=ee64c0d09794e86969c8bf59d36644293bf59f88&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %