4-Bromocinnamic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-(4-Bromophenyl)acrylic acid
B1690-25G
B1690-5G
TCIAB1690-25GEA
1050
NOK
TCIAB1690-25G
TCIAB1690-5G
4-Bromocinnamic acid ≥98.0% (by GC, titration analysis)
4-Bromocinnamic acid
Formel:
C₉H₇BrO₂ MW: 227,06 g/mol |
MDL nummer:
MFCD00004394 CAS-nummer: 1200-07-3 EINECS: 214-850-7 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Pyridine | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...