4-Fluorocinnamic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: (2E)-3-(4-fluorophenyl)prop-2-enoic acid, p-Fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, trans-4-Fluorocinnamic acid

F0244-25G F0244-5G
TCIAF0244-25GEA 875 NOK
TCIAF0244-25G TCIAF0244-5G
4-Fluorocinnamic acid ≥98.0% (by GC, titration analysis)
4-Fluorocinnamic acid
Formel: C₉H₇FO₂
MW: 166,15 g/mol
Smeltepunkt: 209 °C
Tetthet: 1 g/cm³ (20 °C)
MDL nummer: MFCD00004395
CAS-nummer: 459-32-5
EINECS: 207-288-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5adf12edc826c7606c91fb88b8b58c858a60371&vid=a5adf12edc826c7606c91fb88b8b58c858a60371&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 207.0 to 211.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR