(±)-3-(Diethylamino)-1,2-propanediol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: (±)-3-(Diethylamino)-1,2-dihydroxy propane, 3-(Diethylamino)propane-1,2-diol

D1721-25G D1721-500G
TCIAD1721-25GEA 650 NOK
TCIAD1721-25G TCIAD1721-500G
(±)-3-(Diethylamino)-1,2-propanediol ≥98.0% (by GC, titration analysis)
(±)-3-(Diethylamino)-1,2-propanediol
Formel: C₇H₁₇NO₂
MW: 147,22 g/mol
Kokepunkt: 233°C
Smeltepunkt: –30 °C
Tetthet: 0,9762 g/cm³ (20 °C)
Flash punkt: 107°C
MDL nummer: MFCD00004716
CAS-nummer: 621-56-7
EINECS: 210-693-3
FN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2abca30fc11bcd06d987e6c21c7e87ac051406eb&vid=2abca30fc11bcd06d987e6c21c7e87ac051406eb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR