(±)-1,2-Butanediol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1,2-Dihydroxybutane, (±)-1,2-Butylene glycol, (±)-1,2-Dihydroxy butane, 1,2-Butylene glycol, Butane-1,2-diol, 1,2-Butanediol

B0804-500G B0804-25G
TCIAB0804-500GEA 610 NOK
TCIAB0804-500G TCIAB0804-25G
(±)-1,2-Butanediol ≥98.0% (by GC)
(±)-1,2-Butanediol
Formel: C₄H₁₀O₂
MW: 90,12 g/mol
Kokepunkt: 193 °C (1013 hPa)
Smeltepunkt: –50 °C
Tetthet: 1,0189 g/cm³ (0 °C)
MDL nummer: MFCD00004570
CAS-nummer: 584-03-2
EINECS: 209-527-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dff144a2140af5c179af439cbf9b0908458d42b&vid=7dff144a2140af5c179af439cbf9b0908458d42b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR